Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ints9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253731.1 | NP_001240660.1 | 1 | 310 | ||
NM_153414.4 | NP_700463.2 | 1 | 310 | |||
GenBank mRNA | AK035769.1 | - | 1 | 54 | ||
AK038979.1 | - | 1 | 43 | |||
AK077634.1 | - | 1 | 311 | |||
AK136992.1 | - | 1 | 37 | |||
AK150195.1 | - | 1 | 37 | |||
BY195822.1 | - | 1 | 43 |