Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153534.2 | NP_705762.2 | 3 | 712 | ||
GenBank mRNA | AK122432.1 | - | 3 | 527 | ||
AK132312.1 | - | 3 | 713 | |||
BC037107.1 | - | 3 | 469 | 5367175 |