Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pygb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153781.1 | NP_722476.1 | 11 | 1453 | ||
GenBank mRNA | AK032769.1 | - | 11 | 1466 | ||
AK035673.1 | - | 11 | 1183 | |||
AK133794.1 | - | 2 | 183 | |||
AK134667.1 | - | 11 | 1463 | |||
AK142716.1 | - | 11 | 1463 | |||
AK147763.1 | - | 11 | 1485 | |||
AK168381.1 | - | 11 | 1464 | |||
AK169052.1 | - | 11 | 1488 | |||
AK169631.1 | - | 11 | 1467 | |||
AK182124.1 | - | 1 | 61 | |||
AK213064.1 | - | 1 | 116 | |||
BC031457.1 | - | 11 | 1461 | |||
BC032209.1 | - | 5 | 608 | |||
BC035283.1 | - | 11 | 1453 | 4947455 |