Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170599.2 | NP_733548.2 | 7 | 1473 | ||
GenBank mRNA | AK160146.1 | - | 7 | 1436 | ||
BC057555.1 | - | 7 | 1473 | 6416190 |