Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047433.2 | NP_001040898.1 | 2 | 322 | ||
NM_001284346.1 | NP_001271275.1 | 3 | 397 | |||
NM_172254.4 | NP_758458.1 | 3 | 397 | |||
GenBank mRNA | AK011448.1 | - | 3 | 315 | ||
AK017764.1 | - | 3 | 441 | |||
AK048097.1 | - | 3 | 287 | |||
BC029910.1 | - | 3 | 288 | 4920779 | ||
BC039954.1 | - | 3 | 308 | 4193739 | ||
BG082470.2 | - | 2 | 336 | |||
BY257422.1 | - | 2 | 420 | |||
DN175457.1 | - | 2 | 296 |