Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142288.1 | NP_001135760.1 | 4 | 1517 | ||
NM_006067.4 | NP_006058.1 | 5 | 1612 | |||
XM_017022867.1 | XP_016878356.1 | 5 | 1303 | |||
GenBank mRNA | BC001472.2 | - | 5 | 1562 | 3138396 | |
BC005886.2 | - | 5 | 1562 | 3546377 | ||
BC007445.1 | - | 5 | 1553 | 3009805 | ||
BC020250.1 | - | 5 | 1562 | 4653699 |