Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thoc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172438.3 | NP_766026.1 | 4 | 364 | ||
GenBank mRNA | AK050734.1 | - | 4 | 364 | ||
AK173078.1 | - | 2 | 202 | |||
AK207100.1 | - | 1 | 30 | |||
BC020332.1 | - | 3 | 329 | |||
BC021779.1 | - | 3 | 338 | |||
BC039758.1 | - | 4 | 365 | 3995737 |