Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Inpp5j | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172439.3 | NP_766027.2 | 6 | 1782 | ||
GenBank mRNA | AK034272.1 | - | 6 | 1783 | ||
AK136908.1 | - | 6 | 1769 | |||
AK165718.1 | - | 5 | 2310 | |||
AK170114.1 | - | 6 | 1748 | |||
AK171029.1 | - | 6 | 1743 | |||
BC022915.1 | - | 6 | 1735 | |||
BC046561.1 | - | 5 | 595 | |||
BC051452.1 | - | 6 | 1751 | |||
BC131634.1 | - | 6 | 1777 | 40096940 | ||
BC131635.1 | - | 6 | 1774 | 40096943 |