Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172447.2 | NP_766035.1 | 18 | 2275 | ||
GenBank mRNA | AK029553.1 | - | 18 | 2283 | ||
AK039313.1 | - | 18 | 2276 | |||
AK076835.1 | - | 18 | 2265 | |||
AK140211.1 | - | 14 | 1741 | |||
AK164190.1 | - | 10 | 1220 | |||
BC050971.1 | - | 8 | 961 | |||
BC054079.1 | - | 18 | 2226 | |||
BC062906.1 | - | 18 | 2240 | 6813710 |