Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3hav1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172467.3 | NP_766055.1 | 4 | 975 | ||
GenBank mRNA | AK045287.1 | - | 4 | 976 | ||
AK051064.1 | - | 4 | 977 | |||
AK051832.1 | - | 2 | 363 | |||
AK084475.1 | - | 4 | 907 | |||
BC138180.1 | - | 4 | 1067 | 8861199 | ||
BC138181.1 | - | 4 | 1067 | 8861200 |