Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105196.1 | NP_001098666.1 | 6 | 1191 | ||
NM_001105197.1 | NP_001098667.1 | 7 | 982 | |||
NM_001271489.1 | NP_001258418.1 | 7 | 1277 | |||
NM_001271490.1 | NP_001258419.1 | 7 | 1449 | |||
NM_001271491.1 | NP_001258420.1 | 6 | 911 | |||
NM_172472.3 | NP_766060.2 | 7 | 1296 | |||
GenBank mRNA | AK089762.1 | - | 5 | 953 | ||
AK139756.1 | - | 6 | 1188 | |||
AK144657.1 | - | 7 | 1261 | |||
AK144751.1 | - | 7 | 1264 | |||
AK145029.1 | - | 7 | 1417 | |||
AK145832.1 | - | 7 | 1261 | |||
AK147405.1 | - | 5 | 2977 | |||
AK150372.1 | - | 7 | 1048 | |||
AK151447.1 | - | 7 | 967 | |||
AK152221.1 | - | 7 | 967 | |||
AK153051.1 | - | 7 | 1048 | |||
AK154690.1 | - | 6 | 1847 | |||
AK155883.1 | - | 6 | 913 | |||
AK159162.1 | - | 7 | 1177 | |||
AK220116.1 | - | 4 | 277 | |||
BC056358.1 | - | 7 | 1284 | 6813486 | ||
BC063047.1 | - | 7 | 1284 | 6813486 | ||
S76673.1 | - | 5 | 634 |