Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tfe3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105196.1 | NP_001098666.1 | 9 | 2333 | ||
NM_001105197.1 | NP_001098667.1 | 10 | 2124 | |||
NM_001271489.1 | NP_001258418.1 | 10 | 2419 | |||
NM_001271490.1 | NP_001258419.1 | 10 | 2591 | |||
NM_001271491.1 | NP_001258420.1 | 9 | 2053 | |||
NM_172472.3 | NP_766060.2 | 10 | 2438 | |||
GenBank mRNA | AK089762.1 | - | 8 | 2095 | ||
AK139756.1 | - | 9 | 2330 | |||
AK144657.1 | - | 10 | 2403 | |||
AK144751.1 | - | 10 | 2408 | |||
AK145029.1 | - | 10 | 2560 | |||
AK145832.1 | - | 10 | 2404 | |||
AK147405.1 | - | 7 | 5497 | |||
AK150372.1 | - | 10 | 2190 | |||
AK151447.1 | - | 10 | 2109 | |||
AK152221.1 | - | 10 | 2109 | |||
AK153051.1 | - | 10 | 2190 | |||
AK154690.1 | - | 8 | 3893 | |||
AK159162.1 | - | 10 | 2318 | |||
BC056358.1 | - | 10 | 2426 | 6813486 | ||
BC063047.1 | - | 10 | 2426 | 6813486 | ||
S76673.1 | - | 8 | 1776 |