Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hace1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172473.3 | NP_766061.2 | 3 | 454 | ||
GenBank mRNA | AK042879.1 | - | 3 | 395 | ||
AK156958.1 | - | 3 | 396 | |||
BC120695.1 | - | 3 | 351 | 40129618 | ||
BC120697.1 | - | 3 | 351 | 40129620 |