Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tyw3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168358.1 | NP_001161830.1 | 5 | 1679 | ||
NM_172474.5 | NP_766062.1 | 6 | 1813 | |||
GenBank mRNA | AK028611.1 | - | 1 | 48 | ||
AK030344.1 | - | 6 | 1811 | |||
AK034820.1 | - | 6 | 1797 |