Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thsd7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172485.3 | NP_766073.2 | 4 | 1433 | ||
GenBank mRNA | AK047508.1 | - | 1219 | |||
AK051714.1 | - | 4 | 1275 | |||
AK083798.1 | - | 4 | 1261 | |||
BC063250.1 | - | 4 | 1263 | 6410178 |