Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lacc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172488.2 | NP_766076.1 | 3 | 809 | ||
GenBank mRNA | AK033565.1 | - | 3 | 810 | ||
AK053693.1 | - | 3 | 1094 | |||
AK152154.1 | - | 3 | 750 | |||
AK153349.1 | - | 3 | 834 | |||
BC116748.1 | - | 2 | 565 | 40126067 | ||
BC119140.1 | - | 2 | 565 | 8733889 |