Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sepsecs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172490.3 | NP_766078.1 | 5 | 631 | ||
GenBank mRNA | AK033654.1 | - | 5 | 632 | ||
AK049338.1 | - | 638 | ||||
AK153792.1 | - | 571 | ||||
BC062133.1 | - | 5 | 621 | 6516578 |