Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006088.5 | NP_006079.1 | 4 | 1085 | ||
GenBank mRNA | AK000560.1 | - | 4 | 1057 | ||
AK021414.1 | - | 4 | 913 | |||
AK026167.1 | - | 4 | 1056 | |||
AK026594.1 | - | 4 | 1057 | |||
AK026609.1 | - | 4 | 1057 | |||
AK098686.1 | - | 4 | 986 | |||
AY167990.1 | - | 4 | 983 | |||
BC001911.1 | - | 4 | 1020 | 3544236 | ||
BC002783.1 | - | 4 | 1030 | 3614023 | ||
BC002885.2 | - | 4 | 1027 | 3944735 | ||
BC004188.2 | - | 4 | 1032 | 2964559 | ||
BC007889.2 | - | 4 | 1021 | 4127195 | ||
BC008006.1 | - | 1 | 331 | |||
BC012835.2 | - | 4 | 1032 | 3603755 | ||
BC019359.1 | - | 4 | 1023 | 4024979 | ||
BC019829.1 | - | 4 | 1046 | 5110038 | ||
BC024038.1 | - | 4 | 1021 | 3616531 | ||
BC029529.1 | - | 4 | 1057 | 5274343 | ||
BC039175.1 | - | 4 | 1030 | 4385885 | ||
BC071889.1 | - | 4 | 1021 | 4905049 | ||
BX648521.1 | - | 3 | 1531 |