Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Boc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172506.2 | NP_766094.1 | 9 | 2149 | ||
GenBank mRNA | AF388037.1 | - | 8 | 1602 | ||
AK028770.1 | - | 9 | 2170 | |||
AK028889.1 | - | 9 | 2150 | |||
AK131182.1 | - | 4 | 4369 | |||
BC062892.1 | - | 9 | 2129 | 6418202 | ||
BC078631.1 | - | 9 | 2093 | 6515418 |