Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171007.1 | NP_001164478.1 | 3 | 561 | ||
NM_172729.3 | NP_766317.1 | 3 | 812 | |||
GenBank mRNA | AK089662.1 | - | 3 | 562 | ||
AK137585.1 | - | 3 | 817 | |||
AY160222.1 | - | 2 | 347 | |||
BC042670.1 | - | 3 | 639 | 3597911 |