Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grm6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173372.2 | NP_775548.2 | 11 | 3388 | ||
GenBank mRNA | BC022720.1 | - | 2 | 1056 | ||
BC034118.1 | - | 7 | 2389 | |||
BC041684.1 | - | 6 | 2397 | 4502870 |
Set Membership: |
![]() |