Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CORO2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190456.1 | NP_001177385.1 | 3 | 288 | ||
NM_001190457.1 | NP_001177386.1 | 3 | 567 | |||
NM_001324014.1 | NP_001310943.1 | 4 | 499 | |||
NM_001324015.1 | NP_001310944.1 | 3 | 300 | |||
NM_006091.4 | NP_006082.3 | 3 | 522 | |||
GenBank mRNA | AB010098.1 | - | 3 | 255 | ||
AB023142.1 | - | 2 | 226 | |||
AK289678.1 | - | 3 | 567 | |||
BC026335.1 | - | 3 | 238 | 4792415 | ||
BX461395.2 | - | 3 | 521 | |||
DA128408.1 | - | 3 | 288 | |||
DA182763.1 | - | 3 | 257 | |||
DA219458.1 | - | 3 | 249 | |||
DA291058.1 | - | 3 | 249 |