Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078167.2 | NP_001071635.1 | 3 | 907 | ||
NM_173374.4 | NP_775550.2 | 3 | 907 | |||
GenBank mRNA | AK018176.1 | - | 3 | 576 | ||
AK053674.1 | - | 3 | 598 | |||
AK078715.1 | - | 3 | 564 | |||
AK150535.1 | - | 3 | 610 | |||
AK169203.1 | - | 3 | 564 | |||
AK169623.1 | - | 3 | 565 | |||
AK187083.1 | - | 2 | 90 | |||
BC046773.1 | - | 3 | 539 | 5708288 | ||
BC058627.1 | - | 3 | 573 | 6401674 | ||
BY743476.1 | - | 3 | 610 | |||
X66091.1 | - | 3 | 520 |