Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001078167.2 | NP_001071635.1 | 3 | 1444 | ||
NM_173374.4 | NP_775550.2 | 4 | 1248 | |||
GenBank mRNA | AK018176.1 | - | 4 | 917 | ||
AK027960.1 | - | 1 | 41 | |||
AK053674.1 | - | 4 | 939 | |||
AK150535.1 | - | 4 | 951 | |||
AK169203.1 | - | 4 | 905 | |||
AK169623.1 | - | 4 | 906 | |||
BC046773.1 | - | 4 | 880 | 5708288 | ||
BC058627.1 | - | 4 | 914 | 6401674 | ||
X66091.1 | - | 3 | 1057 |