Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbmx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173376.3 | NP_775552.1 | 5 | 450 | ||
GenBank mRNA | AK076115.1 | - | 5 | 451 | ||
AK160232.1 | - | 5 | 378 | |||
AK168018.1 | - | 5 | 365 | |||
BC026976.1 | - | 5 | 375 | 5361950 |