Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbmx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173376.3 | NP_775552.1 | 6 | 1500 | ||
GenBank mRNA | AK076115.1 | - | 6 | 1501 | ||
AK168018.1 | - | 6 | 1415 | |||
BC026976.1 | - | 6 | 1425 | 5361950 |