Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pamr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173749.4 | NP_776110.3 | 2 | 259 | ||
GenBank mRNA | AK031254.1 | - | 2 | 132 | ||
AK088017.1 | - | 2 | 158 | |||
BB620582.1 | - | 2 | 126 | |||
BC031841.1 | - | 2 | 213 | 3675412 | ||
BC057685.1 | - | 2 | 213 | 3675412 |