Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pamr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC031841.1 | - | 4 | 561 | 3675412 | |
BC057685.1 | - | 4 | 561 | 3675412 |