Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174991.3 | NP_778156.2 | 2 | 1262 | ||
GenBank mRNA | AK147456.1 | - | 2 | 1247 | ||
AK147459.1 | - | 2 | 1215 | |||
AK147494.1 | - | 2 | 1049 | |||
AK147600.1 | - | 2 | 1024 | |||
AK147607.1 | - | 2 | 1266 | |||
AK220481.1 | - | 2 | 627 | |||
AY168408.1 | - | 2 | 774 | |||
BC115835.1 | - | 2 | 768 |
Set Membership: |
G-protein coupled receptor |