Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174991.3 | NP_778156.2 | 28 | 4657 | ||
GenBank mRNA | AK046831.1 | - | 12 | 1175 | ||
AK147456.1 | - | 28 | 4642 | |||
AK147459.1 | - | 28 | 4610 | |||
AK147494.1 | - | 28 | 4444 | |||
AK147600.1 | - | 27 | 4333 | |||
AK147607.1 | - | 28 | 4661 | |||
AK220481.1 | - | 28 | 4051 | |||
AY168408.1 | - | 28 | 4169 | |||
BC036963.1 | - | 10 | 835 | |||
BC059857.1 | - | 7 | 587 |
Set Membership: |
G-protein coupled receptor |