Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adgrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174991.3 | NP_778156.2 | 15 | 3366 | ||
GenBank mRNA | AK147456.1 | - | 15 | 3351 | ||
AK147459.1 | - | 15 | 3319 | |||
AK147494.1 | - | 15 | 3153 | |||
AK147600.1 | - | 15 | 3128 | |||
AK220481.1 | - | 15 | 2745 | |||
AY168408.1 | - | 15 | 2878 |
Set Membership: |
G-protein coupled receptor |