Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175360.2 | NP_780569.1 | 9, 10 | 1210 | ||
GenBank mRNA | AK009962.1 | - | 2, 3 | 129 | ||
AK075574.1 | - | 9, 10 | 1210 | |||
AK146929.1 | - | 9, 10 | 1288 | |||
AK164386.1 | - | 9, 10 | 1220 | |||
BC029548.1 | - | 9, 10 | 1250 | 5012064 |