Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4933402P03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175368.1 | NP_780577.1 | 3 | 581 | ||
GenBank mRNA | AK077090.1 | - | 3 | 609 | ||
BC137961.1 | - | 3 | 570 | 8860980 |