Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175369.3 | NP_780578.1 | 6 | 1563 | ||
GenBank mRNA | AK077161.1 | - | 6 | 1564 | ||
AK136053.1 | - | 2 | 810 |