Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Als2cr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175370.4 | NP_780579.1 | 2, 3 | 242 | ||
GenBank mRNA | AK016678.1 | - | 2, 3 | 400 | ||
AK077190.1 | - | 2, 3 | 210 | |||
AK133373.1 | - | 241 | ||||
BC145039.1 | - | 1, 2 | 151 | |||
BC145040.1 | - | 1, 2 | 151 |