Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtrf1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175374.3 | NP_780583.1 | 3 | 629 | ||
GenBank mRNA | AK036424.1 | - | 3 | 533 | ||
AK078897.1 | - | 3 | 597 | |||
AK185802.1 | - | 3 | 236 |