Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam72a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175382.3 | NP_780591.1 | 4 | 1150 | ||
GenBank mRNA | AK041641.1 | - | 4 | 1152 | ||
AK076058.1 | - | 4 | 1172 | |||
BC090647.1 | - | 4 | 1236 | 30633261 | ||
BC106123.1 | - | 4 | 1134 | 3995296 | ||
BC138225.1 | - | 4 | 1119 | 8861244 | ||
BC145667.1 | - | 4 | 1119 | 40130970 |