Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198006.4 | NP_932123.3 | 3 | 3206 | ||
GenBank mRNA | AK075747.1 | - | 1 | 1311 | ||
AK166651.1 | - | 3 | 3135 | |||
AK169639.1 | - | 3 | 3193 | |||
BC069969.1 | - | 1 | 1761 | |||
BC096450.1 | - | 1 | 1822 |