Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175402.4 | NP_780611.3 | 1 | 1366 | ||
GenBank mRNA | AK075777.1 | - | 1 | 309 | ||
BC052180.1 | - | 2 | 548 | 6332977 | ||
BC078452.1 | - | 2 | 1189 |