Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175414.4 | NP_780623.1 | 7, 8 | 753 | ||
GenBank mRNA | AK079139.1 | - | 7, 8 | 726 | ||
AK158729.1 | - | 6, 7 | 638 | |||
AK163395.1 | - | 7, 8 | 809 | |||
AK203288.1 | - | 1, 2 | 99 | |||
BC052503.1 | - | 7, 8 | 735 | 5146854 |