Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adtrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145875.1 | NP_001139347.1 | 2 | 238 | ||
NM_175417.4 | NP_780626.1 | 2 | 159 | |||
GenBank mRNA | AK029025.1 | - | 2 | 161 | ||
AK079178.1 | - | 2 | 239 | |||
AK135327.1 | - | 2 | 239 | |||
BC027755.1 | - | 1 | 68 | 5055136 | ||
BY719442.1 | - | 2 | 235 |