Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adtrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145875.1 | NP_001139347.1 | 4 | 594 | ||
NM_175417.4 | NP_780626.1 | 4 | 515 | |||
GenBank mRNA | AK029025.1 | - | 4 | 517 | ||
AK079178.1 | - | 4 | 596 | |||
BC027755.1 | - | 3 | 424 | 5055136 | ||
BY719442.1 | - | 4 | 591 |