Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135172.1 | NP_001128644.1 | 3 | 1598 | ||
NM_175425.4 | NP_780634.2 | 3 | 1241 | |||
GenBank mRNA | AK017441.1 | - | 3 | 1590 | ||
AK078818.1 | - | 3 | 1404 | |||
BC038308.1 | - | 3 | 1241 | |||
BC090967.1 | - | 3 | 1579 | 30652983 | ||
DQ002400.1 | - | 3 | 1250 |