Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175429.3 | NP_780638.1 | 3 | 589 | ||
GenBank mRNA | AK046557.1 | - | 3 | 478 | ||
AK046708.1 | - | 3 | 516 | |||
AK050097.1 | - | 2 | 451 | |||
AK085035.1 | - | 3 | 517 | |||
BC064459.1 | - | 3 | 589 | 6821736 |