Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grik4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175481.5 | NP_780690.2 | 11 | 1280 | ||
GenBank mRNA | AK032029.1 | - | 11 | 1281 | ||
AK080582.1 | - | 11 | 1408 | |||
BC118010.2 | - | 10 | 1195 | 40096690 |