Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ltbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113549.1 | NP_001107021.1 | 4, 5 | 841 | ||
NM_175641.2 | NP_783572.2 | 7, 8 | 1040 | |||
GenBank mRNA | AF410798.1 | - | 7, 8 | 1121 | ||
AF410799.1 | - | 4, 5 | 817 | |||
AK054297.1 | - | 4, 5 | 807 | |||
AK147753.1 | - | 4, 5 | 842 | |||
AK157031.1 | - | 4, 5 | 857 | |||
BC032073.1 | - | 4, 5 | 829 | |||
BC059016.1 | - | 4, 5 | 818 | |||
BC141005.1 | - | 7, 8 | 1046 | 9055617 |