Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006137.6 | NP_006128.1 | 2 | 399 | ||
XM_011523608.1 | XP_011521910.1 | 2 | 366 | |||
XM_017025316.1 | XP_016880805.1 | 2 | 432 | |||
GenBank mRNA | AK223465.1 | - | 2 | 371 | ||
AK298096.1 | - | 2 | 324 | |||
AK310061.1 | - | 2 | 326 | |||
AK310558.1 | - | 2 | 372 | |||
AY935534.1 | - | 2 | 371 | |||
AY935535.1 | - | 2 | 371 | |||
AY935536.1 | - | 2 | 371 | |||
AY935537.1 | - | 2 | 371 | |||
BC009293.2 | - | 2 | 343 | 4120480 | ||
BC013297.2 | - | 2 | 343 | 3529600 | ||
BI838777.1 | - | 2 | 400 | |||
BT006696.1 | - | 2 | 290 | |||
X06180.1 | - | 1 | 777 |