Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A530053G22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK079908.1 | - | 4 | 507 | ||
AK080052.1 | - | 5 | 605 | |||
BC051224.1 | - | 4 | 521 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177329.1 | - | 604 | Chr. - - [-] on Build |