Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ghsr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177330.4 | NP_796304.1 | 3 | 1798 | ||
GenBank mRNA | AK049671.1 | - | 3 | 1799 |
Set Membership: |
G-protein coupled receptor |