Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159658.1 | NP_001153130.1 | 1 | 490 | ||
NM_177587.2 | NP_808255.1 | 1 | 490 | |||
GenBank mRNA | AB084104.1 | - | 1 | 553 | ||
AK131815.1 | - | 1 | 491 | |||
BC107212.1 | - | 1 | 484 | 40051607 | ||
BC107213.1 | - | 1 | 482 | 40051613 |